missing translation for 'onlineSavingsMsg'
Learn More

2-[(4-Chlorophenyl)thio]benzaldehyde, 95%, Thermo Scientific™

Product Code. 10059061
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10059061 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10059061 Supplier Thermo Scientific Maybridge Supplier No. KM02217DA
 Restricted Item
Estimated Shipment: 31-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 107572-07-6
CAS Min % 95.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Molecular Formula C13H9ClOS
Linear Formula ClC6H4SC6H4CHO
MDL Number MFCD00051766
Quantity 1 g
Synonym 2-4-chlorophenylthio benzaldehyde, 2-4-chlorophenyl thio benzaldehyde, 2-4-chlorophenyl sulfanyl benzaldehyde, 2-4-chlorophenyl sulfanylbenzaldehyde, benzaldehyde,2-4-chlorophenyl thio, acmc-20aopf, 2-4-chlorophenyl sulphanyl benzaldehyde
InChI Key CQKLAEUCMKGSEQ-UHFFFAOYSA-N
SMILES C1=CC=C(C(=C1)C=O)SC2=CC=C(C=C2)Cl
IUPAC Name 2-(4-chlorophenyl)sulfanylbenzaldehyde
Molecular Weight (g/mol) 248.724
PubChem CID 2778624
Formula Weight 248.73
Percent Purity 95%
Chemical Name or Material 2-[(4-Chlorophenyl)thio]benzaldehyde
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.