missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Chlorosulfonylphenyl)ethyltrimethoxysilane, 50% solution in dichloromethane, AcroSeal™

Product Code. 10321820 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10321820 100 mL AcroSeal™ Glass Bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10321820 Supplier Thermo Scientific Acros Supplier No. 428991000
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 126519-89-9 | C11H17ClO5SSi | 324.85 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 126519-89-9, 75-09-2
CAS Min % 45.0
CAS Max % 55.0
Color Yellow
Boiling Point >40.0°C
Infrared Spectrum Authentic
Packaging AcroSeal™ Glass Bottle
Molecular Formula C11H17ClO5SSi
MDL Number MFCD00054774
Quantity 100 mL
Synonym 2-4-chlorosulfonylphenyl ethyltrimethoxysilane, 4-2-trimethoxysilylethyl benzenesulfonyl chloride, 4-2-trimethoxysilyl ethyl benzene-1-sulfonyl chloride, benzenesulfonylchloride, 4-2-trimethoxysilyl ethyl, 2-4-chlorosulfonylphenyl ethyltrimethoxysilane in methylene chloride, has free sulfonic acid, 4-2-trimethoxysilyl ethyl benzenesulfonyl chloride, acmc-20ete8, 4-chlorosulfonyl phenethyltrimethoxysilane
Solubility Information Solubility in water: reacts
InChI Key NYIDSUMRGUILGR-UHFFFAOYSA-N
SMILES CO[Si](CCC1=CC=C(C=C1)S(Cl)(=O)=O)(OC)OC
IUPAC Name 4-(2-trimethoxysilylethyl)benzenesulfonyl chloride
Molecular Weight (g/mol) 324.85
PubChem CID 2733807
Formula Weight 324.85
Percent Purity 45 to 55% (solids)
Physical Form Solution
Chemical Name or Material 2-(4-Chlorosulfonylphenyl)ethyltrimethoxysilane
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-(4-Chlorosulfonylphenyl)ethyltrimethoxysilane, 50% solution in dichloromethane
Signal Word
  • Danger
Hazard Category
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H351-Suspected of causing cancer.
  • H336-May cause drowsiness or dizziness.
  • H314-Causes severe skin burns and eye damage.
  • EUH029-Contact with water liberates toxic gas.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.