missing translation for 'onlineSavingsMsg'
Learn More

2,4-Diamino-6-hydroxy-5-nitrosopyrimidine, 97%, Thermo Scientific™

Product Code. 11499412
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11499412 100 g 100g
11489412 25 g 25g
11409422 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11499412 Supplier Thermo Scientific Alfa Aesar Supplier No. A10147.22
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2387-48-6
Molecular Formula C4H5N5O2
Molecular Weight (g/mol) 155.117
MDL Number MFCD00023263
InChI Key HVMRLFSFHWCUCG-UHFFFAOYSA-N
Synonym 2,4-diamino-6-hydroxy-5-nitrosopyrimidine, 2,6-diamino-5-nitrosopyrimidin-4-ol, 4-pyrimidinol, 2,6-diamino-5-nitroso, 4 1h-pyrimidinone, 2,6-diamino-5-nitroso, 2,6-diamino-5-nitroso-4-pyrimidinol, 4-pyrimidol, 2,6-diamino-5-nitroso, 2,6-diamino-5-nitrosopyrimidin-4 3h-one, d04tlb, acmc-1cg71, 4-pyrimidol,6-diamino-5-nitroso
PubChem CID 75436
IUPAC Name 2,6-diamino-5-nitroso-1H-pyrimidin-4-one
SMILES C1(=C(NC(=NC1=O)N)N)N=O

Specifications

Melting Point >360°C
Quantity 100 g
Formula Weight 155.12
Percent Purity 97%
Chemical Name or Material 2,4-Diamino-6-hydroxy-5-nitrosopyrimidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.