missing translation for 'onlineSavingsMsg'
Learn More

2,4-Diamino-6-piperidinopyrimidine (Desoxyminoxidil), TRC

Product Code. 30007673
Change view
Click to view available options
Quantity:
100 mg
250 mg
500 mg
Unit Size:
100mg
250mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30007673 250 mg 250mg
30003627 500 mg 500mg
17982585 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30007673 Supplier Toronto Research Chemicals Supplier No. TRCD416640250MG
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24867-26-3
Molecular Formula C9 H15 N5
Molecular Weight (g/mol) 193.25
Synonym 6-(Piperidin-1-yl)pyrimidine-2,4-diamine, Minoxidil Imp. E (EP), Desoxyminoxidil
IUPAC Name 6-piperidin-1-ylpyrimidine-2,4-diamine
SMILES Nc1cc(nc(N)n1)N2CCCCC2

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C9H15N5/c10-7-6-8(13-9(11)12-7)14-4-2-1-3-5-14/h6H,1-5H2,(H4,10,11,12,13)
Formula Weight 193.1327
Percent Purity >95%
Chemical Name or Material 6-(Piperidin-1-yl)pyrimidine-2,4-diamine (Desoxyminoxidil)
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.