missing translation for 'onlineSavingsMsg'
Learn More

2,4-Diaminotoluene, 98%

Product Code. 11465497
Change view
Click to view available options
Quantity:
250 g
1000 g
Unit Size:
1000g
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11465497 250 g 250g
11455497 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11465497 Supplier Thermo Scientific Alfa Aesar Supplier No. B23495.30
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 95-80-7 | C7H10N2 | 122.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 95-80-7
Molecular Formula C7H10N2
Molecular Weight (g/mol) 122.17
MDL Number MFCD00007804
InChI Key VOZKAJLKRJDJLL-UHFFFAOYSA-N
Synonym 2,4-diaminotoluene, 2,4-toluenediamine, developer t, toluene-2,4-diamine, 1,3-benzenediamine, 4-methyl, developer mtd, m-toluenediamine, fourrine m, m-tolylenediamine, benzofur mt
PubChem CID 7261
ChEBI CHEBI:34237
IUPAC Name 4-methylbenzene-1,3-diamine
SMILES CC1=CC=C(N)C=C1N

Specifications

Melting Point 96°C to 101°C
Boiling Point 283°C to 285°C
Flash Point 169°C (336°F)
Odor Recognizable
Quantity 250 g
UN Number UN1709
Beilstein 2205839
Formula Weight 122.17
Percent Purity 98%
Chemical Name or Material 2,4-Diaminotoluene
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 2,4-Diaminotoluene
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Carcinogenicity Category 1B
  • Germ cell mutagenicity Category 2
  • Reproductive toxicity Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H301-Toxic if swallowed.
  • H312-Harmful in contact with skin.
  • H317-May cause an allergic skin reaction.
  • H341-Suspected of causing genetic defects.
  • H350-May cause cancer.
  • H361f-Suspected of damaging fertility.
  • H373-May cause damage to organs.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: Toluene 2,4-diamine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.