missing translation for 'onlineSavingsMsg'
Learn More

2,4-Diaminotoluene, TRC

Product Code. 30353753
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
Product Code. Quantity unitSize
30353753 10 g 10g
1 options
This item is not returnable. View return policy

Product Code. 30353753

Brand: Toronto Research Chemicals TRCD41678010G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2,4-Diaminotoluene
CAS 95-80-7
Molecular Formula C7 H10 N2
Quantity 10 g
Synonym Toluene-2,4-diamine (7CI,8CI), 4-Methyl-1,3-benzenediamine, 1,3-Diamino-4-methylbenzene, 2,4-Diamino-1-methylbenzene, 2,4-Diaminotoluene, 2,4-Tolylenediamine, 2-Methyl-1,5-benzenediamine, 3-Amino-4-methylaniline, 4-Methyl-1,3-diaminobenzene, 4-Methyl-1,3-phenylenediamine, 4-Methyl-m-phenylenediamine, Brown for Fur T, C.I. Oxidation Base 35, Eucanine GB, Fouramine J, Fourrine 94, Fourrine M, Nako TMT, Pelagol Grey J, Pelagol J, Pontamine Developer TN, Renal MD, TDA, TDA (diamine), Zoba GKE, m-Toluenediamine, m-Tolylenediamine
InChI Formula InChI=1S/C7H10N2/c1-5-2-3-6(8)4-7(5)9/h2-4H,8-9H2,1H3
SMILES Cc1ccc(N)cc1N
IUPAC Name 4-methylbenzene-1,3-diamine
Molecular Weight (g/mol) 122.17
Formula Weight 122.0844
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.