missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dibromo-3-pentanone, mixture of stereoisomers, 97%

Product Code. 11452668
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11452668 10 g 10g
11462668 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11452668 Supplier Thermo Scientific Alfa Aesar Supplier No. A16680.09
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 815-60-1 | C5H8Br2O | 243.926 g/mol

2,4-Dibromo-2,4-dimethyl-3-pentanone may be used in the synthesis of 2-dimethylamino-4-methylene-1,3-dioxolanes, via debromination using zinc-copper couple in dimethylformamide and dimethylacetamide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4-Dibromo-2,4-dimethyl-3-pentanone may be used in the synthesis of 2-dimethylamino-4-methylene-1,3-dioxolanes, via debromination using zinc-copper couple in dimethylformamide and dimethylacetamide.

Solubility
Slightly soluble in water(50 g/L (20°C).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, reducing agents, bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 815-60-1
Molecular Formula C5H8Br2O
Molecular Weight (g/mol) 243.926
MDL Number MFCD00015719
InChI Key UOPIOAUZQKSZRO-UHFFFAOYSA-N
Synonym 2,4-dibromo-3-pentanone, 2,4-dibromo-pentan-3-one, 3-pentanone,2,4-dibromo, 2,4-bis bromanyl pentan-3-one, 2,4-dibromo-3-pentanone, mixture of stereoisomers
PubChem CID 11791395
IUPAC Name 2,4-dibromopentan-3-one
SMILES CC(C(=O)C(C)Br)Br

Specifications

Boiling Point 67°C to 82°C (10 mmHg)
Odor Pungent
Quantity 10 g
UN Number UN3265
Solubility Information Slightly soluble in water(50g/L (20°C).
Formula Weight 243.91
Percent Purity 97%
Chemical Name or Material 2,4-Dibromo-3-pentanone, mixture of stereoisomers

RUO – Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.