missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dibromobenzenesulfonyl chloride, 98%

Product Code. 11379905
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11379905 5 g 5g
11369905 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11379905 Supplier Thermo Scientific Alfa Aesar Supplier No. L19887.06
 Restricted Item
Estimated Shipment: 14-10-2026
Add to basket
This item is not returnable. View return policy

CAS: 72256-95-2 | C6H3Br2ClO2S | 334.41 g/mol

2,4-Dibromobenzenesulfonyl chloride is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4-Dibromobenzenesulfonyl chloride is used as pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Moisture sensitive. Incompatible with oxidizing agents and bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 72256-95-2
Molecular Formula C6H3Br2ClO2S
Molecular Weight (g/mol) 334.41
MDL Number MFCD03093759
InChI Key OMOXIKAWVFDFNX-UHFFFAOYSA-N
Synonym 2,4-dibromobenzene-1-sulfonyl chloride, 2,4-dibromobenzenesulphonyl chloride, 2,4-dibromophenyl chlorosulfone, pubchem5111, 2,4-dibromophenylsulfonyl chloride, benzenesulfonylchloride, 2,4-dibromo, 2,4-bis bromanyl benzenesulfonyl chloride, 2,4-dibromobenzenesulfonyl chloride
PubChem CID 2737366
IUPAC Name 2,4-dibromobenzenesulfonyl chloride
SMILES ClS(=O)(=O)C1=C(Br)C=C(Br)C=C1

Specifications

Melting Point 82°C to 86°C
Boiling Point 128°C (0.2 mmHg)
Quantity 5 g
UN Number UN3261
Beilstein 2694058
Sensitivity Moisture sensitive
Solubility Information Insoluble in water.
Formula Weight 334.41
Percent Purity 98%
Chemical Name or Material 2,4-Dibromobenzenesulfonyl chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.