missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dibromothiazole, 97%

Product Code. 15406708
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15406708 1 g 1g
15416708 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15406708 Supplier Thermo Scientific Alfa Aesar Supplier No. H60230.03
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 4175-77-3 | C3HBr2NS | 242.916 g/mol

2,4-Dibromothiazole is applied as a building block in a synthesis of Melithiazole C employing a highly (E)-selective cross-metathesis between the derived 4-vinylthiazole and a dienote side-chain. It is also used as an intermediate in the preparation of various 2,4-disubstituted thiazole derivatives with biologically active properties such as survival motor neuron (SMN) protein modulators.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4-Dibromothiazole is applied as a building block in a synthesis of Melithiazole C employing a highly (E)-selective cross-metathesis between the derived 4-vinylthiazole and a dienote side-chain. It is also used as an intermediate in the preparation of various 2,4-disubstituted thiazole derivatives with biologically active properties such as survival motor neuron (SMN) protein modulators.

Notes
Air sensitive. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4175-77-3
Molecular Formula C3HBr2NS
Molecular Weight (g/mol) 242.916
MDL Number MFCD01318994
InChI Key MKEJZKKVVUZXIS-UHFFFAOYSA-N
Synonym 2,4-dibromothiazole, 2,4-dibromo thiazole, dibromothiazole, 2.4-dibromothiazole, soxlllqacqdfwmijp@, 2,4-dibromo-thiazole, acmc-209jll, ksc235o5d, 2 pound not4-dibromo-thiazole
PubChem CID 2763297
IUPAC Name 2,4-dibromo-1,3-thiazole
SMILES C1=C(N=C(S1)Br)Br

Specifications

Melting Point 80°C to 84°C
Quantity 1 g
Formula Weight 242.92
Percent Purity 97%
Chemical Name or Material 2,4-Dibromothiazole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.