missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dibromothiophene, 97%, Thermo Scientific™

Product Code. 10403023
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10403023 1 g 1g
10239974 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10403023 Supplier Thermo Scientific Maybridge Supplier No. CC28510DA

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 3140-92-9
Molecular Formula C4H2Br2S
Molecular Weight (g/mol) 241.928
MDL Number MFCD00043889
InChI Key WAQFYSJKIRRXLP-UHFFFAOYSA-N
Synonym thiophene, 2,4-dibromo, 2,4 dibromothiophene, pubchem1908, 2,4-dibromo-thiophene, acmc-1cq64
PubChem CID 2724560
IUPAC Name 2,4-dibromothiophene
SMILES C1=C(SC=C1Br)Br

Specifications

CAS Min % 94.5
CAS Max % 100.0
Density 2.1400g/mL
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 241.93
Percent Purity 95%
Chemical Name or Material 2, 4-Dibromothiophene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.