missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dichloro-5-nitropyrimidine, TRC

Product Code. 30376951
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30376951 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30376951 Supplier Toronto Research Chemicals Supplier No. TRCD4356401G
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2,4-Dichloro-5-nitropyrimidine
CAS 49845-33-2
Molecular Formula C4 H Cl2 N3 O2
Quantity 1 g
InChI Formula InChI=1S/C4HCl2N3O2/c5-3-2(9(10)11)1-7-4(6)8-3/h1H
SMILES [O-][N+](=O)c1cnc(Cl)nc1Cl
IUPAC Name 2,4-dichloro-5-nitropyrimidine
Molecular Weight (g/mol) 193.98
Formula Weight 192.9446
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.