missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dichloro-6-methylpyrimidine, 98%

Product Code. 11494163
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11494163 100 g 100g
11474163 5 g 5g
11484163 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11494163 Supplier Thermo Scientific Alfa Aesar Supplier No. A10363.22
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 5424-21-5 | C5H4Cl2N2 | 163.001 g/mol

2,4-Dichloro-6-methylpyrimidine is used in the synthesis of (2-chloro-6-methyl-pyrimidin-4-yl)-(2,3-dihydro-benzothiazol-6-yl)-amine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4-Dichloro-6-methylpyrimidine is used in the synthesis of (2-chloro-6-methyl-pyrimidin-4-yl)-(2,3-dihydro-benzothiazol-6-yl)-amine.

Solubility
Soluble in Chloroform, Ether, Ethyl Acetate and Toluene. Insoluble in water.

Notes
Store away from oxidizing agents and bases. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5424-21-5
Molecular Formula C5H4Cl2N2
Molecular Weight (g/mol) 163.001
MDL Number MFCD00006064
InChI Key BTLKROSJMNFSQZ-UHFFFAOYSA-N
Synonym pyrimidine, 2,4-dichloro-6-methyl, 2,6-dichloro-4-methylpyrimidine, 2,4-dichloro-6-methyl-pyrimidine, 2,4-dichloro-6-methyl pyrimidine, 6-methyl-2,4-dichloropyrimidine, 2,4-dichlor-6-methylpyrimidin, pubchem6911, acmc-1b11m, ksc495i8r, 2,4-dichloro6-methylpyrimidine
PubChem CID 79471
IUPAC Name 2,4-dichloro-6-methylpyrimidine
SMILES CC1=CC(=NC(=N1)Cl)Cl

Specifications

Melting Point 44°C to 47°C
Boiling Point 219°C
Flash Point >110°C (230°F)
Quantity 100 g
UN Number UN1759
Beilstein 114295
Solubility Information Soluble in Chloroform,Ether,Ethyl Acetate and Toluene. Insoluble in water.
Formula Weight 163.01
Percent Purity 98%
Chemical Name or Material 2,4-Dichloro-6-methylpyrimidine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.