missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dichloro-6-methylpyrimidine, TRC

Product Code. 30376205
Change view
Click to view available options
Quantity:
200 g
Unit Size:
200g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30376205 200 g 200g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30376205 Supplier Toronto Research Chemicals Supplier No. TRCD435505200G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2,4-Dichloro-6-methylpyrimidine
CAS 5424-21-5
Molecular Formula C5 H4 Cl2 N2
Quantity 200 g
InChI Formula InChI=1S/C5H4Cl2N2/c1-3-2-4(6)9-5(7)8-3/h2H,1H3
SMILES Cc1cc(Cl)nc(Cl)n1
IUPAC Name 2,4-dichloro-6-methylpyrimidine
Molecular Weight (g/mol) 163
Formula Weight 161.9752
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.