missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dichloro-L-phenylalanine, 98%, Thermo Scientific™

Product Code. 11416480
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11416480 1 g 1g
15473717 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11416480 Supplier Thermo Scientific Alfa Aesar Supplier No. H27579.03
 Restricted Item
Estimated Shipment: 04-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111119-36-9
Molecular Formula C9H9Cl2NO2
Molecular Weight (g/mol) 234.08
MDL Number MFCD01860882
InChI Key GWHQTNKPTXDNRM-QMMMGPOBSA-N
Synonym 2,4-dichloro-l-phenylalanine, l-2,4-dichlorophenylalanine, 2s-2-amino-3-2,4-dichlorophenyl propanoic acid, s-2-amino-3-2,4-dichlorophenyl propanoic acid, h-phe 2,4-dicl-oh, l-2,4-dichlorophe, l-phenylalanine, 2,4-dichloro, h-phe 2,4-cl2-oh, 2,4-dichloro-phenylalanine, s-2-amino-3-2,4-dichloro-phenyl-propionic acid
PubChem CID 2761497
IUPAC Name (2S)-2-amino-3-(2,4-dichlorophenyl)propanoic acid
SMILES N[C@@H](CC1=CC=C(Cl)C=C1Cl)C(O)=O

Spezifikation

Quantity 1 g
Formula Weight 234.09
Percent Purity 98%
Chemical Name or Material 2,4-Dichloro-L-phenylalanine
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.