missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dichloro-N-hydroxybenzenecarboximidoyl chloride, 97%, Thermo Scientific™

Product Code. 10651503
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10651503 10 g Amber Glass Bottle 10g
10466292 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10651503 Supplier Thermo Scientific Maybridge Supplier No. SPB06594EA
 Restricted Item
Estimated Shipment: 04-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 29203-60-9
Molecular Formula C7H4Cl3NO
Molecular Weight (g/mol) 224.465
MDL Number MFCD00192643
InChI Key GXBVZCSCOBTMHS-XFFZJAGNSA-N
Synonym 2,4-dichloro-n-hydroxybenzenecarboximidoyl chloride, 2,4-dichloro-n-hydroxybenzenecarbonimidoyl chloride, 2,4-dichloro-n-hydroxybenzimidoyl chloride, 2,4-dichlorophenyl chloro hydroxyimino methane, 2,4-dichloro-n-hydroxybenzene-carboximidoyl chloride, z-2,4-dichloro-n-hydroxybenzene-1-carbonimidoyl chloride
PubChem CID 9583124
IUPAC Name (1Z)-2,4-dichloro-N-hydroxybenzenecarboximidoyl chloride
SMILES C1=CC(=C(C=C1Cl)Cl)C(=NO)Cl

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Linear Formula Cl2C6H3CCl=NOH
Quantity 10 g
Formula Weight 224.47
Percent Purity 97%
Chemical Name or Material 2, 4-Dichloro-N-hydroxybenzenecarboximidoyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.