missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dichlorobenzaldehyde oxime, 97%, Thermo Scientific™

Product Code. 10579782
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10579782 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10579782 Supplier Thermo Scientific Maybridge Supplier No. SPB04865DA
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 56843-28-8
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C7H5Cl2NO
Linear Formula Cl2C6H3CH=NOH
MDL Number MFCD00173694
Quantity 1 g
Synonym 2,4-dichlorobenzaldehyde oxime, 2,4-dichlorobenzaldoxime, benzaldoxime,4-dichloro, wln: qnu1r bg dg, 2,4-dichorobenzaldehyde oxime, benzaldehyde,4-dichloro-, oxime, 2,4-dichlorophenyl hydroxyimino methane, e-n-2,4-dichlorophenyl methylidene hydroxylamine, ne-n-2,4-dichlorophenyl methylidene hydroxylamine, z-n-2,4-dichlorophenyl methylidene hydroxylamine
InChI Key ONJQBRVMFRQQIG-ONNFQVAWSA-N
SMILES O\N=C\C1=CC=C(Cl)C=C1Cl
IUPAC Name (E)-N-[(2,4-dichlorophenyl)methylidene]hydroxylamine
Molecular Weight (g/mol) 190.02
PubChem CID 6434542
Formula Weight 190.03
Percent Purity 97%
Chemical Name or Material 2, 4-Dichlorobenzaldehyde oxime
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.