missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dichlorobenzoyl Chloride, TRC

Product Code. 30369256
Change view
Click to view available options
Quantity:
250 g
Unit Size:
250g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30369256 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30369256 Supplier Toronto Research Chemicals Supplier No. TRCD431715250G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2,4-Dichlorobenzoyl Chloride
CAS 89-75-8
Molecular Formula C7H3Cl3O
Quantity 250 g
Synonym o,p-Dichlorobenzoyl Chloride
InChI Formula InChI=1S/C7H3Cl3O/c8-4-1-2-5(7(10)11)6(9)3-4/h1-3H
SMILES ClC(C1=CC=C(Cl)C=C1Cl)=O
IUPAC Name 2,4-dichlorobenzoyl chloride
Molecular Weight (g/mol) 209.46
Formula Weight 207.92
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.