missing translation for 'onlineSavingsMsg'
Learn More

(2,4-Dichlorophenyl)-methanesulfonyl chloride, Thermo Scientific™

Product Code. 10473360
Change view
Click to view available options
Quantity:
5 g
Packaging:
Amber Glass Bottle
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10473360 5 g Amber Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10473360 Supplier Thermo Scientific Maybridge Supplier No. MAY00259DE
 Restricted Item
Estimated Shipment: 06-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 88691-50-3
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C7H5Cl3O2S
Quantity 5 g
Synonym 2,4-dichlorophenyl methanesulfonyl chloride, 2,4-dichlorophenyl-methanesulfonyl chloride, 2,4-dichlorobenzylsulfonyl chloride, 2,4-dichlorophenyl methylsulphonyl chloride, 2,4-dichlorobenzylsulphonyl chloride, benzenemethanesulfonylchloride, 2,4-dichloro, acmc-20do05, 2,4-dichlorophenyl methyl chlorosulfone
InChI Key SNLHLLYEHFIELA-UHFFFAOYSA-N
SMILES C1=CC(=C(C=C1Cl)Cl)CS(=O)(=O)Cl
IUPAC Name (2,4-dichlorophenyl)methanesulfonyl chloride
Molecular Weight (g/mol) 259.525
PubChem CID 594471
Formula Weight 259.53
Percent Purity 97%
Chemical Name or Material (2, 4-Dichlorophenyl)-methanesulfonyl chloride
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.