missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dichlorophenylacetic acid, 99%, Thermo Scientific™

Product Code. 10735801 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10735801 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10735801 Supplier Thermo Scientific Acros Supplier No. 134270100

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 19719-28-9
CAS Min % 98.5
CAS Max % 100.0
Melting Point 128.0°C to 133.0°C
Color Beige-Yellow to White
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Molecular Formula C8H6Cl2O2
Linear Formula Cl2C6H3CH2COOH
MDL Number MFCD00004318
Quantity 10 g
Beilstein 09, III, 2271
Synonym 2,4-dichlorophenylacetic acid, 2-2,4-dichlorophenyl acetic acid, benzeneacetic acid, 2,4-dichloro, 2,4-dichloro-phenyl-acetic acid, 2,4-dichlorophenyl acetic acid, 2 ,4-dichlorophenylacetic acid, zlchem 477, pubchem15026, acmc-209f0x
Solubility Information Solubility in water: soluble in hot water. Other solubilities: soluble in ether,alcohol and chloroform
InChI Key GXMWLJKTGBZMBH-UHFFFAOYSA-N
SMILES C1=CC(=C(C=C1Cl)Cl)CC(=O)O
IUPAC Name 2-(2,4-dichlorophenyl)acetic acid
Molecular Weight (g/mol) 205.04
PubChem CID 88209
Formula Weight 205.04
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 2, 4-Dichlorophenylacetic acid, 99%
Show More Show Less
compliance-icons
Product Identifier
  • 2,4-Dichlorophenylacetic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.