missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dichlorophenylboronic acid, 97%, Thermo Scientific Chemicals

Product Code. 10733413 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10733413 1 g Glass bottle 1g
10601853 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10733413 Supplier Thermo Scientific Acros Supplier No. 367670010
 Restricted Item
Estimated Shipment: 18-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 68716-47-2 | Molecular Formula: C6H5BCl2O2 | Molecular Weight: 190.81 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68716-47-2
Molecular Formula C6H5BCl2O2
Molecular Weight (g/mol) 190.81
MDL Number MFCD00013930
InChI Key QNEGDGPAXKYZHZ-UHFFFAOYSA-N
Synonym 2,4-dichlorophenyl boronic acid, 2,4-dichlorobenzeneboronic acid, 2,4-dichlorophenylbornic acid, 2,4-dichlorophenyl boranediol, chembl20726, 2,4-dichloro benzene boronic acid, boronic acid, 2,4-dichlorophenyl, 2,4-dichlorobenzeneboron dihydroxide, 2,4-dichlorobenzeneboronicacid
PubChem CID 2734659
IUPAC Name (2,4-dichlorophenyl)boronic acid
SMILES OB(O)C1=CC=C(Cl)C=C1Cl

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 240.0°C to 245.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Quantity 1 g
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in methanol
Formula Weight 190.82
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 2, 4-Dichlorophenylboronic acid, 97%
Show More Show Less
compliance-icons
Product Identifier
  • 2,4-Dichlorophenylboronic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P264-Wash face, hands and any thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.