missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dichloropyridine, 97%

Product Code. 11394446
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11394446 1 g 1g
11304456 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11394446 Supplier Thermo Scientific Alfa Aesar Supplier No. L20040.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 26452-80-2 | C5H3Cl2N | 147.986 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26452-80-2
Molecular Formula C5H3Cl2N
Molecular Weight (g/mol) 147.986
MDL Number MFCD00955618
InChI Key TYPVHTOETJVYIV-UHFFFAOYSA-N
Synonym pyridine, 2,4-dichloro, 2,4-dichloro pyridine, 2,4-dichoropyridine, 2, 4-dichloropyridine, 2,4-dichloro-pyridine, 2,4-dichlorpyridin, 2,4-dichlorpyridine, 2,4-dichloropyrdine, pubchem3531, acmc-1cnvt
PubChem CID 33515
IUPAC Name 2,4-dichloropyridine
SMILES C1=CN=C(C=C1Cl)Cl

Specifications

Melting Point -1°C
Density 1.37
Boiling Point 189°C to 190°C
Flash Point 89°C (192°F)
Refractive Index 1.5485
Quantity 1 g
UN Number UN2810
Beilstein 108666
Formula Weight 147.99
Percent Purity 97%
Chemical Name or Material 2,4-Dichloropyridine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.