missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dichloropyridine, 98%

Product Code. 10646673 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
Unit Size:
100g
1g
25g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10646673 5 g 5g
10168422 1 g 1g
10398613 25 g 25g
11956331 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10646673 Supplier Thermo Scientific Acros Supplier No. 379470050
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 26452-80-2 | C5H3Cl2N | 147.99 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26452-80-2
Molecular Formula C5H3Cl2N
Molecular Weight (g/mol) 147.99
MDL Number MFCD00955618
InChI Key TYPVHTOETJVYIV-UHFFFAOYSA-N
Synonym pyridine, 2,4-dichloro, 2,4-dichloro pyridine, 2,4-dichoropyridine, 2, 4-dichloropyridine, 2,4-dichloro-pyridine, 2,4-dichlorpyridin, 2,4-dichlorpyridine, 2,4-dichloropyrdine, pubchem3531, acmc-1cnvt
PubChem CID 33515
IUPAC Name 2,4-dichloropyridine
SMILES C1=CN=C(C=C1Cl)Cl

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point -1.0°C
Density 1.3700g/mL
Boiling Point 189.0°C to 190.0°C
Flash Point 89°C
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5500 to 1.5540
Quantity 5 g
Specific Gravity 1.37
Formula Weight 147.99
Percent Purity 98%
Chemical Name or Material 2, 4-Dichloropyridine
Show More Show Less
compliance-icons
Product Identifier
  • 2,4-Dichloropyridine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.