missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dichloropyrimidine, 98+%

Product Code. 11414090
Change view
Click to view available options
Quantity:
5 g
10 g
50 g
250 g
Unit Size:
10g
250g
50g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11414090 5 g 5g
11492098 10 g 10g
11402108 50 g 50g
11412108 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11414090 Supplier Thermo Scientific Alfa Aesar Supplier No. A15131.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 3934-20-1 | C4H2Cl2N2 | 148.974 g/mol

2,4-Dichloropyrimidine is used in the synthesis of medicinally important 4-aryl-5-pyrimidinylimidazoles. It undergoes effective one-pot, regioselective double Suzuki coupling reaction to yield diarylated pyrimidines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4-Dichloropyrimidine is used in the synthesis of medicinally important 4-aryl-5-pyrimidinylimidazoles. It undergoes effective one-pot, regioselective double Suzuki coupling reaction to yield diarylated pyrimidines.

Solubility
Soluble in water (partly), methanol, chloroform, and ethyl acetate.

Notes
Moisture Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3934-20-1
Molecular Formula C4H2Cl2N2
Molecular Weight (g/mol) 148.974
MDL Number MFCD00006061
InChI Key BTTNYQZNBZNDOR-UHFFFAOYSA-N
PubChem CID 77531
IUPAC Name 2,4-dichloropyrimidine
SMILES C1=CN=C(N=C1Cl)Cl

Specifications

Melting Point 57°C to 62°C
Boiling Point 101°C (23 mmHg)
Quantity 5 g
Beilstein 110911
Sensitivity Moisture sensitive
Synonym pyrimidine, 2,4-dichloro, 2,6-dichloropyrimidine, 2,4-dichloro-pyrimidine, 2,4-dichlorpyrimidin, 2,4-dichloro pyrimidine, pyrimidine,2,4-dichloro, pyrimidine,4-dichloro, hydrochloride, zlchem 210, 2,4dichloropyrimidine
Solubility Information Soluble in water (partly),methanol,chloroform,and ethyl acetate.
Formula Weight 148.98
Percent Purity ≥98%
Chemical Name or Material 2,4-Dichloropyrimidine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.