missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dichlorotoluene, 99%

Product Code. 10697132 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mL
1 L
Packaging:
Glass bottle
Unit Size:
1 L
250mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10697132 250 mL Glass bottle 250mL
10123750 1 L Glass bottle 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10697132 Supplier Thermo Scientific Acros Supplier No. 163282500
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 95-73-8 | C7H6Cl2 | 161.03 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 95-73-8
Molecular Formula C7H6Cl2
Molecular Weight (g/mol) 161.03
MDL Number MFCD00000583
InChI Key FUNUTBJJKQIVSY-UHFFFAOYSA-N
Synonym 2,4-dichlorotoluene, benzene, 2,4-dichloro-1-methyl, toluene, 2,4-dichloro, 2,4-dichloro-1-methyl-benzene, unii-3s32n6lg3x, 2,4-dichloromethylbenzene, 2,4-dct, 2,4-diclorotoluene, 4,6-dichlorotoluene, 2,4-dichloro-toluen
PubChem CID 7254
ChEBI CHEBI:81651
IUPAC Name 2,4-dichloro-1-methylbenzene
SMILES CC1=C(C=C(C=C1)Cl)Cl

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -14.0°C
Color Colorless
Density 1.2400g/mL
Boiling Point 200.0°C
Flash Point 79°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3C6H3Cl2
Refractive Index 1.5450 to 1.5470
Quantity 250 mL
Beilstein 05, 295
Specific Gravity 1.24
Solubility Information Solubility in water: immiscible. Other solubilities: soluble in alcohol,ether and acetone
Formula Weight 161.03
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2, 4-Dichlorotoluene, 99%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2,4-Dichlorotoluene
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.