missing translation for 'onlineSavingsMsg'
Learn More

2,4-Difluoro-alpha-(1H-1,2,4-triazolyl)acetophenone, TRC

Product Code. 30136286
Change view
Click to view available options
Quantity:
1 g
5 g
250 g
Unit Size:
1g
250g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30136286 250 g 250g
30132808 5 g 5g
17939454 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136286 Supplier Toronto Research Chemicals Supplier No. TRCD445885250G
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 86404-63-9
Molecular Formula C10 H7 F2 N3 O
Molecular Weight (g/mol) 223.18
Synonym Ethanone, 1-(2,4-difluorophenyl)-2-(1H-1,2,4-triazol-1-yl)-, 1-(2,4-Difluorophenyl)-2-(1H-1,2,4-triazol-1-yl)ethanone, 1-(2,4-Difluorophenyl)-2-(1H-1,2,4-triazol-1-yl)-1-ethanone, 1-(2,4-Difluorophenyl)-2-(4H-1,3,4-triazol-4-yl)ethan-1-one, 2',4'-Difluoro-2-(1,2,4-triazol-1-yl)acetophenone, 2',4'-Difluoro-2-(1H-1,2,4-triazol-1-yl)acetophenone, UK 51060, Voriconazole Imp. A (EP)
IUPAC Name 1-(2,4-difluorophenyl)-2-(1,2,4-triazol-1-yl)ethanone
SMILES Fc1ccc(C(=O)Cn2cncn2)c(F)c1

Specifications

Quantity 250 g
InChI Formula InChI=1S/C10H7F2N3O/c11-7-1-2-8(9(12)3-7)10(16)4-15-6-13-5-14-15/h1-3,5-6H,4H2
Formula Weight 223.0557
Chemical Name or Material 1-(2,4-Difluorophenyl)-2-(1H-1,2,4-triazol-1-yl)ethanone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.