missing translation for 'onlineSavingsMsg'
Learn More

2,4-Difluorobenzenesulfonyl chloride, 98%

Product Code. 11497586
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11497586 5 g 5g
11407596 25 g 25g
11417596 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11497586 Supplier Thermo Scientific Alfa Aesar Supplier No. A14639.06
 Restricted Item
Estimated Shipment: 04-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 13918-92-8 | C6H3ClF2O2S | 212.595 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13918-92-8
Molecular Formula C6H3ClF2O2S
Molecular Weight (g/mol) 212.595
MDL Number MFCD00042481
InChI Key FJSAJUXIHJIAMD-UHFFFAOYSA-N
Synonym 2,4-difluorobenzene-1-sulfonyl chloride, 2,4-difluorobenzenesulphonyl chloride, 2,4-difluoro-benzenesulfonyl chloride, benzenesulfonyl chloride, 2,4-difluoro, 2,4-difluorophenyl chlorosulfone, buttpark 23\07-79, 2,4-difluorophenylsulfonyl chloride, pubchem5122, acmc-209civ
PubChem CID 2734273
IUPAC Name 2,4-difluorobenzenesulfonyl chloride
SMILES C1=CC(=C(C=C1F)F)S(=O)(=O)Cl

Specifications

Density 1.581
Boiling Point 236°C to 237°C
Flash Point >110°C (230°F)
Refractive Index 1.5187
Quantity 5 g
UN Number UN3265
Beilstein 2970373
Sensitivity Moisture sensitive
Formula Weight 212.6
Percent Purity 98%
Chemical Name or Material 2,4-Difluorobenzenesulfonyl chloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.