missing translation for 'onlineSavingsMsg'
Learn More

2',4'-Dihydroxy-3'-methylacetophenone, 98%

Product Code. 11459766
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11459766 25 g 25g
11449766 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11459766 Supplier Thermo Scientific Alfa Aesar Supplier No. B20272.14
 Restricted Item
Estimated Shipment: 21-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 10139-84-1 | C9H10O3 | 166.176 g/mol

chemo selective benzylation of 2,4-dihydroxy-3-methylacetophenone gave the 4-benzyloxy derivative. 4- Ethyl-6 methyl resorcinol was prepared by Clemmensen reduction of 4-dihydroxy-3- methylacetophenone.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
chemo selective benzylation of 2,4-dihydroxy-3-methylacetophenone gave the 4-benzyloxy derivative. 4- Ethyl-6 methyl resorcinol was prepared by Clemmensen reduction of 4-dihydroxy-3- methylacetophenone.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10139-84-1
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.176
MDL Number MFCD00010817
InChI Key KMTLZBUHQPQFAV-UHFFFAOYSA-N
Synonym 1-2,4-dihydroxy-3-methylphenyl ethanone, 2',4'-dihydroxy-3'-methylacetophenone, 1-2,4-dihydroxy-3-methylphenyl ethan-1-one, 2,4-dihydroxy-3-methylacetophenone, ethanone, 1-2,4-dihydroxy-3-methylphenyl, acmc-20aocy, 1-acetyl-2,4-dihydroxy-3-methylbenzene, 2 ',4 '-dihydroxy-3 '-methylacetophenone, 1-2,4-dihydroxy-3-methyl-phenyl ethanone, 1-2,4-dihydroxy-3-methyl-phenyl-ethanone
PubChem CID 592139
IUPAC Name 1-(2,4-dihydroxy-3-methylphenyl)ethanone
SMILES CC1=C(C=CC(=C1O)C(=O)C)O

Specifications

Melting Point 152°C to 156°C
Quantity 25 g
Formula Weight 166.18
Percent Purity 98%
Chemical Name or Material 2',4'-Dihydroxy-3'-methylacetophenone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.