missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dihydroxybenzaldehyde, 98%

Product Code. 11494080
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11494080 500 g 500g
11465293 25 g 25g
11475293 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11494080 Supplier Thermo Scientific Alfa Aesar Supplier No. A12868.36
 Restricted Item
Estimated Shipment: 30-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 95-01-2 | C7H6O3 | 138.12 g/mol

2,4-Dihydroxybenzaldehyde is used in two-step synthesis of ethyl 3,5-dibromo-2,4-dihydroxycinnamate. It has a potent antioxidative and antibacterial activity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4-Dihydroxybenzaldehyde is used in two-step synthesis of ethyl 3,5-dibromo-2,4-dihydroxycinnamate. It has a potent antioxidative and antibacterial activity.

Solubility
Soluble in water. Solubility in methanol is almost transparent. Easily soluble in alcohol, ethyl ether, chloroform, glacial acetic acid. Insoluble in benzene and easily decomposes under strong acid strong alkali.

Notes
Air sensitive. Store away from oxidizing agents and air. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 95-01-2
Molecular Formula C7H6O3
Molecular Weight (g/mol) 138.12
MDL Number MFCD00011686
InChI Key IUNJCFABHJZSKB-UHFFFAOYSA-N
Synonym beta-resorcylaldehyde, 4-hydroxysalicylaldehyde, 4-formylresorcinol, beta-resorcaldehyde, benzaldehyde, 2,4-dihydroxy, beta-resorcinaldehyde, beta-resorcylic aldehyde, 2,4-dihydroxybenzenecarbonal, salicylaldehyde, 4-hydroxy, 4-hydroxysalicyladehyde
PubChem CID 7213
ChEBI CHEBI:50198
IUPAC Name 2,4-dihydroxybenzaldehyde
SMILES OC1=CC=C(C=O)C(O)=C1

Specifications

Melting Point 134°C to 138°C
Boiling Point 220°C (22 mmHg)
Quantity 500 g
Beilstein 878548
Sensitivity Air Sensitive
Merck Index 14,8156
Solubility Information Soluble in water. Solubility in methanol is almost transparent. Easily soluble in alcohol,ethyl ether,chloroform,glacial acetic acid. Insoluble in benzene and easily decomposes under strong acid strong alkali.
Formula Weight 138.12
Percent Purity 98%
Chemical Name or Material 2,4-Dihydroxybenzaldehyde

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.