missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dihydroxypyrimidine-5-carboxylic acid, 98%

Product Code. 10467180 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10467180 5 g Glass bottle 5g
10477180 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10467180 Supplier Thermo Scientific Acros Supplier No. 155200050
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 23945-44-0 | C5H4N2O4 | 156.1 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23945-44-0
Molecular Formula C5H4N2O4
Molecular Weight (g/mol) 156.1
MDL Number MFCD00149397
InChI Key ZXYAAVBXHKCJJB-UHFFFAOYSA-N
Synonym 2,4-dihydroxypyrimidine-5-carboxylic acid, uracil 5-carboxylic acid, isoorotic acid, uracil-5-carboxylic acid, 5-carboxyuracil, 2,4-dioxo-1,2,3,4-tetrahydropyrimidine-5-carboxylic acid, 2,4-dihydroxy-5-pyrimidinecarboxylic acid, 5-uracilcarboxylic acid, 5-pyrimidinecarboxylic acid, 1,2,3,4-tetrahydro-2,4-dioxo, uracil 5-carboxylate
PubChem CID 90301
ChEBI CHEBI:17477
IUPAC Name 2,4-dioxo-1H-pyrimidine-5-carboxylic acid
SMILES C1=C(C(=O)NC(=O)N1)C(=O)O

Especificaciones

CAS Min % 97.5
CAS Max % 100.0
Melting Point 283.0°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Beilstein 25, 256
Formula Weight 156.1
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 2, 4-Dihydroxypyrimidine-5-carboxylic acid, 98%
Mostrar más Mostrar menos

RUO – Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.