missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dihydroxypyrimidine-5-carboxylic acid, 98%

Product Code. 10477180 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10477180 25 g Glass bottle 25g
10467180 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10477180 Supplier Thermo Scientific Acros Supplier No. 155200250
 Restricted Item
Estimated Shipment: 07-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 23945-44-0 | C5H4N2O4 | 156.1 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23945-44-0
Molecular Formula C5H4N2O4
Molecular Weight (g/mol) 156.1
MDL Number MFCD00149397
InChI Key ZXYAAVBXHKCJJB-UHFFFAOYSA-N
Synonym 2,4-dihydroxypyrimidine-5-carboxylic acid, uracil 5-carboxylic acid, isoorotic acid, uracil-5-carboxylic acid, 5-carboxyuracil, 2,4-dioxo-1,2,3,4-tetrahydropyrimidine-5-carboxylic acid, 2,4-dihydroxy-5-pyrimidinecarboxylic acid, 5-uracilcarboxylic acid, 5-pyrimidinecarboxylic acid, 1,2,3,4-tetrahydro-2,4-dioxo, uracil 5-carboxylate
PubChem CID 90301
ChEBI CHEBI:17477
IUPAC Name 2,4-dioxo-1H-pyrimidine-5-carboxylic acid
SMILES C1=C(C(=O)NC(=O)N1)C(=O)O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 283.0°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 25 g
Beilstein 25, 256
Formula Weight 156.1
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 2, 4-Dihydroxypyrimidine-5-carboxylic acid, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.