missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dimethoxybenzaldehyde, 97%, Thermo Scientific™

Product Code. 10116782
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10116782 10 g 10g
10055272 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10116782 Supplier Thermo Scientific Maybridge Supplier No. SB00910EA

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 613-45-6
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.176
MDL Number MFCD00003311
InChI Key LWRSYTXEQUUTKW-UHFFFAOYSA-N
Synonym benzaldehyde, 2,4-dimethoxy, 2,4-dimethoxybenzaldehyd, 2,4-dimethoxy-benzaldehyde, 2,4-bis methyloxy benzaldehyde, ameba resin, pubchem8220, acmc-209mro, 2,4dimethoxybenzaldehyde, argopore tm-mb-cho, 2,4-dimethoxybenzaldehye
PubChem CID 69175
IUPAC Name 2,4-dimethoxybenzaldehyde
SMILES COC1=CC(=C(C=C1)C=O)OC

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Linear Formula (CH3O)2C6H3CHO
Quantity 10 g
Formula Weight 166.18
Percent Purity 97%
Chemical Name or Material 2, 4-Dimethoxybenzaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.