missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dimethoxybenzeneboronic acid, 98%

Product Code. 11485807
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11485807 25 g 25g
11465807 1 g 1g
11475807 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11485807 Supplier Thermo Scientific Alfa Aesar Supplier No. B24374.14
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 133730-34-4 | C8H11BO4 | 181.982 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 133730-34-4
Molecular Formula C8H11BO4
Molecular Weight (g/mol) 181.982
MDL Number MFCD01074590
InChI Key SQTUYFKNCCBFRR-UHFFFAOYSA-N
Synonym 2,4-dimethoxybenzeneboronic acid, 2,4-dimethoxyphenyl boronic acid, 2,4-dimethoxyphenyl boranediol, boronic acid, 2,4-dimethoxyphenyl, 2,4-dimethoxybenzene boronic acid, boronic acid, b-2,4-dimethoxyphenyl, pubchem1823, acmc-209btd, ksc174i6j, 2,4-dimethoxyphenylboronicacid
PubChem CID 2734341
IUPAC Name (2,4-dimethoxyphenyl)boronic acid
SMILES B(C1=C(C=C(C=C1)OC)OC)(O)O

Specifications

Melting Point 115°C to 120°C
Quantity 25 g
Beilstein 4744213
Formula Weight 181.98
Percent Purity 98%
Chemical Name or Material 2,4-Dimethoxybenzeneboronic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.