missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dimethyl-3-ethylpyrrole, 96%

Product Code. 10326650 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10326650 25 g 25g
10102530 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10326650 Supplier Thermo Scientific Acros Supplier No. 116210250
 Restricted Item
Estimated Shipment: 24-11-2026
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 517-22-6 | C8H13N | 123.2 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 517-22-6
MDL Number MFCD00005222
InChI Key ZEBBLOXDLGIMEG-UHFFFAOYSA-N
Synonym 2,4-dimethyl-3-ethylpyrrole, kryptopyrrole, kryptopyrrol, 3-ethyl-2,4-dimethylpyrrole, cryptopyrrole, 2,4-dimethyl-3-ethyl-1h-pyrrole, 1h-pyrrole, 3-ethyl-2,4-dimethyl, pyrrole, 3-ethyl-2,4-dimethyl, acmc-209kvh, 5-20-05-00099 beilstein handbook reference
PubChem CID 10600
IUPAC Name 3-ethyl-2,4-dimethyl-1H-pyrrole
SMILES CCC1=C(NC=C1C)C

Specifications

CAS Min % 95.0
CAS Max % 100.0
Density 0.9130g/mL
Boiling Point 197°C (710.0 mmHg)
Flash Point 71°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Molecular Formula C8H13N
Refractive Index 1.4960 to 1.498
Quantity 25 g
Beilstein 20, I, 47
Specific Gravity 0.913
Molecular Weight (g/mol) 123.2
Formula Weight 123.2
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material 2, 4-Dimethyl-3-ethylpyrrole, 96%
Show More Show Less
compliance-icons
Product Identifier
  • 2,4-Dimethyl-3-ethylpyrrole
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.