missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dimethyl-3-furoyl chloride, 90%, Thermo Scientific™

Product Code. 10250700
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10250700 250 mg Amber Glass Bottle 250mg
10411440 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10250700 Supplier Thermo Scientific Maybridge Supplier No. CC65802CB
 Restricted Item
Estimated Shipment: 12-10-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 15139-39-6
Molecular Formula C7H7ClO2
Molecular Weight (g/mol) 158.581
InChI Key NOEQXZGBSMGWCT-UHFFFAOYSA-N
Synonym 2,4-dimethyl-3-furoyl chloride, 2,4-dimethylfuran-3-carboxylic chloride, 3-furancarbonyl chloride, 2,4-dimethyl-9ci
PubChem CID 13019985
IUPAC Name 2,4-dimethylfuran-3-carbonyl chloride
SMILES CC1=COC(=C1C(=O)Cl)C

Specifications

Assay Percent Range 90%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 158.58
Percent Purity 90%
Chemical Name or Material 2, 4-Dimethyl-3-furoyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.