missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dimethyl-5-oxazolecarboxylic Acid, TRC

Product Code. 30380679
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30380679 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30380679 Supplier Toronto Research Chemicals Supplier No. TRCD476330500MG
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2,4-Dimethyl-5-oxazolecarboxylic Acid
CAS 2510-37-4
Molecular Formula C6H7NO3
Quantity 500 mg
Synonym 2,4-Dimethyl-1,3-oxazole-5-carboxylic Acid, 2,4-Dimethyloxazole-5-carboxylic Acid, 5-Carboxy-2,4-dimethyloxazole
InChI Formula InChI=1S/C6H7NO3/c1-3-5(6(8)9)10-4(2)7-3/h1-2H3,(H,8,9)
SMILES Cc1oc(C(=O)O)c(C)n1
IUPAC Name 2,4-dimethyl-1,3-oxazole-5-carboxylic acid
Molecular Weight (g/mol) 141.12
Formula Weight 141.0426
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.