missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dimethyl-6-hydroxypyrimidine, 99+%, Thermo Scientific™

Product Code. 10419410 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10419410 5 g Glass bottle 5g
10284090 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10419410 Supplier Thermo Scientific Acros Supplier No. 116350050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6622-92-0
Molecular Formula C6H8N2O
Molecular Weight (g/mol) 124.14
MDL Number MFCD00006105
InChI Key UQFHLJKWYIJISA-UHFFFAOYSA-N
Synonym 2,4-dimethyl-6-hydroxypyrimidine, 2,6-dimethylpyrimidin-4-ol, 4-hydroxy-2,6-dimethylpyrimidine, 6-hydroxy-2,4-dimethylpyrimidine, 2,6-dimethyl-4-pyrimidinol, 4 1h-pyrimidinone, 2,6-dimethyl, 2,6-dimethyl-4-hydroxypyrimidine, 2,6-dimethylpyrimidin-4 3h-one, 2,4-dimethyl-6-oxypyrimidine, t6vm dnj c1 e1
PubChem CID 81098
IUPAC Name 2,6-dimethyl-1H-pyrimidin-4-one
SMILES CC1=CC(=O)N=C(N1)C

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point 198.5°C to 200°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Quantity 5 g
Solubility Information Solubility in water: soluble in water. Other solubilities: soluble in methanol and ethanol,slightly soluble in acetone,insoluble in hydrocarbons
Formula Weight 124.14
Percent Purity 99+%
Physical Form Crystalline Needles
Chemical Name or Material 2, 4-Dimethyl-6-hydroxypyrimidine, 99+%
Show More Show Less

RUO – Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.