missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2,4-Dimethylphenol, 98%

Product Code. 11453984
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11453984 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11453984 Supplier Alfa Aesar™ Supplier No. A11966.22
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Applications
It is used as a perfuming agent in cosmetic industry. Also used in the production of high-viscosity phosphate esters, as a feedstock for hindered phenol antioxidant and specialty modified phenolic resin manufacture. Also used as disinfectant and sanitizer on hospital premises.

Solubility
Slightly soluble in water 4068 mg/L @ 25°C (est).

Notes
Store away from oxidizing agents. Store under nitrogen.
TRUSTED_SUSTAINABILITY

Specifications

Melting Point 21°C to 23°C
CAS 105-67-9
Quantity 100 g
Molecular Formula C8H10O
MDL Number MFCD00002233
UN Number UN2261
Beilstein 636244
Merck Index 14,10082
Synonym 2,4-xylenol, phenol, 2,4-dimethyl, gallex, 4,6-dimethylphenol, 1-hydroxy-2,4-dimethylbenzene, 4-hydroxy-1,3-dimethylbenzene, rcra waste number u101, 2,4-dimethyl phenol, caswell no. 907a, lysol brand disinfectant
Boiling Point 210°C to 212°C
InChI Key KUFFULVDNCHOFZ-UHFFFAOYSA-N
SMILES CC1=CC(=C(C=C1)O)C
IUPAC Name 2,4-dimethylphenol
Molecular Weight (g/mol) 122.167
PubChem CID 7771
ChEBI CHEBI:34241
Formula Weight 122.17
Percent Purity 98%
Chemical Name or Material 2,4-Dimethylphenol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.