missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dimethylphenol, 99%

Product Code. 10381681 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10381681 25 g Glass bottle 25g
10314222 5 g Glass bottle 5g
10177420 100 g Glass bottle 100g
10540004 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10381681 Supplier Thermo Scientific Acros Supplier No. 408450250
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 105-67-9 | C8H10O | 122.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 105-67-9
Molecular Formula C8H10O
Molecular Weight (g/mol) 122.17
MDL Number MFCD00002233
InChI Key KUFFULVDNCHOFZ-UHFFFAOYSA-N
Synonym 2,4-xylenol, phenol, 2,4-dimethyl, gallex, 4,6-dimethylphenol, 1-hydroxy-2,4-dimethylbenzene, 4-hydroxy-1,3-dimethylbenzene, rcra waste number u101, 2,4-dimethyl phenol, caswell no. 907a, lysol brand disinfectant
PubChem CID 7771
ChEBI CHEBI:34241
IUPAC Name 2,4-dimethylphenol
SMILES CC1=CC(=C(C=C1)O)C

Specifications

Melting Point 21.0°C to 23.0°C
CAS Min % 98.5
Color Colorless to Yellow
CAS Max % 100.0
Density 1.0270g/mL
Quantity 25 g
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Liquid
Refractive Index 1.5375 to 1.5405
Assay Percent Range 98.5% min. (GC)
Specific Gravity 1.027
Linear Formula (CH3)2C6H3OH
Beilstein 06, 486
Merck Index 15, 10279
Boiling Point 211.0°C
Solubility Information Solubility in water: 0.5% (25°C). Other solubilities: freely soluble in alcohol,chloroform,ether,benz,ene,soluble in naoh solutions
Formula Weight 122.17
Percent Purity 99%
Chemical Name or Material 2, 4-Dimethylphenol
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 2,4-Dimethylphenol
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P330-Rinse mouth.
  • P331-Do NOT induce vomiting.
  • P363-Wash contaminated clothing before reuse.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: 2,4-Dimethylphenol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.