missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dimethylthiazole-5-sulfonyl chloride, 97%

Product Code. 15450838
Change view
Click to view available options
Quantity:
250 mg
5 g
Unit Size:
250mg
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15450838 250 mg 250mg
15440838 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15450838 Supplier Thermo Scientific Alfa Aesar Supplier No. H54578.MD
 Restricted Item
Estimated Shipment: 18-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 80466-80-4 | C5H6ClNO2S2 | 211.678 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 80466-80-4
Molecular Formula C5H6ClNO2S2
Molecular Weight (g/mol) 211.678
MDL Number MFCD00052746
InChI Key GFFJSTHQILQFNQ-UHFFFAOYSA-N
Synonym 2,4-dimethyl-1,3-thiazole-5-sulfonylchloride, 2,4-dimethylthiazole-5-sulfonyl chloride, 2,4-dimethyl-thiazole-5-sulfonyl chloride, dimethyl-1,3-thiazole-5-sulfonyl chloride, 2,4-dimethyl-5-thiazolesulfonyl chloride, 5-thiazolesulfonyl chloride, 2,4-dimethyl, 2,4-dimethyl-1,3-thiazole-5-sulphonyl chloride, 5-thiazolesulfonylchloride, 2,4-dimethyl, 2,4-dimethylthiazol-5-ylsulphonyl chloride
PubChem CID 2776248
IUPAC Name 2,4-dimethyl-1,3-thiazole-5-sulfonyl chloride
SMILES CC1=C(SC(=N1)C)S(=O)(=O)Cl

Specifications

Boiling Point 82°C to 84°C (0.3 mmHg)
Quantity 250 mg
UN Number UN3265
Sensitivity Moisture sensitive
Formula Weight 211.68
Percent Purity 97%
Chemical Name or Material 2,4-Dimethylthiazole-5-sulfonyl chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.