missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dimethylthiazole, 98%, Thermo Scientific™

Product Code. 10172380 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10172380 5 g Glass bottle 5g
10090890 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10172380 Supplier Thermo Scientific Acros Supplier No. 116860050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 541-58-2
Molecular Formula C5H7NS
Molecular Weight (g/mol) 113.18
MDL Number MFCD00014509
InChI Key OBSLLHNATPQFMJ-UHFFFAOYSA-N
Synonym 2,4-dimethylthiazole, thiazole, 2,4-dimethyl, 2,4-methylthiazole, 2,4-dimethyl thiazole, unii-lht97038oa, 2,4-dimethyl-thiazole, dimethylthiazol, zlchem 533, 2,4 dimethylthiazole, 2.4-dimethylthiazole
PubChem CID 10934
IUPAC Name 2,4-dimethyl-1,3-thiazole
SMILES CC1=NC(C)=CS1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Density 1.0500g/mL
Boiling Point 144°C to 146°C (719.0 mmHg)
Flash Point 42°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5081 to 1.5101
Quantity 5 g
Beilstein 27, IV, 981
Merck Index 11, 3249
Specific Gravity 1.05
Solubility Information Solubility in water: miscible with ice-cold water. Other solubilities: soluble in alcohol,ether
Formula Weight 113.18
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2, 4-Dimethylthiazole, 98%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2,4-Dimethylthiazole
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.