missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dinitrobenzenesulfonyl chloride, 98%

Product Code. 11416226
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11416226 25 g 25g
11406226 5 g 5g
11426226 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11416226 Supplier Thermo Scientific Alfa Aesar Supplier No. B23035.14
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1656-44-6 | C6H3ClN2O6S | 266.608 g/mol

2,4-Dinitrobenzenesulfonyl chloride was used to protect primary amines. It was used as starting reagent in the synthesis of tert-butyl 2-[(2,4-dinitrophenyl) sulfonyl]amino acetate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4-Dinitrobenzenesulfonyl chloride was used to protect primary amines. It was used as starting reagent in the synthesis of tert-butyl 2-[(2,4-dinitrophenyl) sulfonyl]amino acetate.

Solubility
Reacts with water.

Notes
Moisture sensitive. Store away from oxidizing agents, bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1656-44-6
Molecular Formula C6H3ClN2O6S
Molecular Weight (g/mol) 266.608
MDL Number MFCD00007431
InChI Key SSFSNKZUKDBPIT-UHFFFAOYSA-N
Synonym 2,4-dinitrobenzene-1-sulfonyl chloride, benzenesulfonyl chloride, 2,4-dinitro, 2,4-dinitrobenzenesulphonyl chloride, benzenesulfonylchloride, 2,4-dinitro, 2,4-dinitrobenzenesulfonylchloride, 2,4-dinitrophenylsulfonyl chloride, zlchem 879, acmc-20a62d, ksc178i6f, 2,4-dinitrobenzene sulfochloride
PubChem CID 74255
IUPAC Name 2,4-dinitrobenzenesulfonyl chloride
SMILES C1=CC(=C(C=C1[N+](=O)[O-])[N+](=O)[O-])S(=O)(=O)Cl

Specifications

Melting Point 100°C to 104°C
Quantity 25 g
UN Number UN3096
Sensitivity Moisture sensitive
Solubility Information Reacts with water.
Formula Weight 266.62
Percent Purity 98%
Chemical Name or Material 2,4-Dinitrobenzenesulfonyl chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.