missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dinitrofluorobenzene, 98%

Product Code. 10784411 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10784411 25 g Glass bottle 25g
10724041 5 g Glass bottle 5g
10605732 100 g Glass bottle 100g
10145443 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10784411 Supplier Thermo Scientific Acros Supplier No. 117030250
ChemsChrom Mega Deals
Use promo code 27993
View all promotions
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 70-34-8 | C6H3FN2O4 | 186.1 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Alkylation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 70-34-8
Molecular Formula C6H3FN2O4
Molecular Weight (g/mol) 186.1
MDL Number MFCD00007056
InChI Key LOTKRQAVGJMPNV-UHFFFAOYSA-N
Synonym 2,4-dinitrofluorobenzene, dinitrofluorobenzene, fdnb, dnfb, 2,4-dinitro-1-fluorobenzene, sanger's reagent, fluoro-2,4-dinitrobenzene, 2,4-dinitrobenzenefluoride, 2,4-dinitrophenyl fluoride, fluorodinitrobenzene
PubChem CID 6264
ChEBI CHEBI:53049
IUPAC Name 1-fluoro-2,4-dinitrobenzene
SMILES C1=CC(=C(C=C1[N+](=O)[O-])[N+](=O)[O-])F

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 23°C to 26°C
Color Brown to Yellow
Density 1.4800g/mL
Boiling Point 296°C
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula (O2N)2C6H3F
Refractive Index 1.5680 to 1.57
Quantity 25 g
Beilstein 05,262
Fieser 01,321; 02,175; 06,233
Merck Index 15,4204
Specific Gravity 1.48
Solubility Information Solubility in water: 400mg/l (25°C). Other solubilities: soluble in ether,benzene,propylene glycol,and hot alcohol
Formula Weight 186.1
Percent Purity 98%
Physical Form Liquid or Low Melting Crystals
Chemical Name or Material 2, 4-Dinitrofluorobenzene
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2,4-Dinitrofluorobenzene
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Carcinogenicity Category 2
  • Respiratory sensitiser Category 1
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H351-Suspected of causing cancer.
  • H373-May cause damage to organs.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.