missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dinitrophenyl thiocyanate, 98%, Thermo Scientific Chemicals

Product Code. 11411167
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11411167 25 g 25g
11421167 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11411167 Supplier Thermo Scientific Alfa Aesar Supplier No. B24147.14
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1594-56-5
Molecular Formula C7H3N3O4S
Molecular Weight (g/mol) 225.178
MDL Number MFCD00047702
InChI Key XQDQRCRASHAZBA-UHFFFAOYSA-N
Synonym dntb, nirit, trirodazeen, gryzbol, grzybol, 2,4-dinitrothiocyanobenzene, nirit supra, tri-rodazene, 2,4-dinitrothiocyanatobenzene, rhodandinitrobenzol
PubChem CID 15325
ChEBI CHEBI:53051
IUPAC Name (2,4-dinitrophenyl) thiocyanate
SMILES C1=CC(=C(C=C1[N+](=O)[O-])[N+](=O)[O-])SC#N

Specifications

Melting Point 138°C to 140°C
Odor Irritating
Quantity 25 g
Beilstein 1983292
Sensitivity Moisture sensitive
Formula Weight 225.18
Percent Purity 98%
Chemical Name or Material 2,4-Dinitrophenyl thiocyanate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.