missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dioxo-1,2,3,4-tetrahydro-5-pyrimidinylboronic acid, 97%, Thermo Scientific™

Product Code. 10748403
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10748403 250 mg Amber Glass Bottle 250mg
10103282 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10748403 Supplier Thermo Scientific Maybridge Supplier No. MO07137CB
 Restricted Item
Estimated Shipment: 19-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 70523-22-7
Molecular Formula C4H5BN2O4
Molecular Weight (g/mol) 155.90
MDL Number MFCD01318983
InChI Key PVEJOCQTIVCDNO-UHFFFAOYSA-N
Synonym uracil-5-boronic acid, 2,4-dioxo-1,2,3,4-tetrahydropyrimidin-5-yl boronic acid, 2,4-dioxo-1,2,3,4-tetrahydro-5-pyrimidinylboronic acid, 2,4-dihydroxypyrimidine-5-boronic acid, 2,4-dioxo-1,2,3,4-tetrahydropyrimidin-5-ylboronic acid, 2,4-dihydroxypyrimidin-5-ylboronic acid, 2,4-dioxo-1h-pyrimidin-5-yl boronic acid, 2,4-dioxo-1,3-dihydropyrimidin-5-ylboronic acid, t6mvmvj ebqq wln, 5-dihydroxyboryluracil
PubChem CID 256205
IUPAC Name (2,4-dioxo-1H-pyrimidin-5-yl)boronic acid
SMILES OB(O)C1=CNC(=O)NC1=O

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 155.9
Percent Purity 97%
Chemical Name or Material 2,4-Dioxo-1,2,3,4-tetrahydro-5-pyrimidinylboronic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.