missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dioxo-1,2,3,4-tetrahydroquinazoline-6-sulfonyl chloride, 97%

Product Code. 15451656
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15451656 250 mg 250mg
15431656 1 g 1g
15441656 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15451656 Supplier Thermo Scientific Alfa Aesar Supplier No. H54833.MD
 Restricted Item
Estimated Shipment: 21-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 56044-12-3 | C8H5ClN2O4S | 260.648 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56044-12-3
Molecular Formula C8H5ClN2O4S
Molecular Weight (g/mol) 260.648
MDL Number MFCD07186482
InChI Key LEFXQSDJQGOMPS-UHFFFAOYSA-N
Synonym 2,4-dioxo-1,2,3,4-tetrahydro-quinazoline-6-sulfonyl chloride, 2,4-dioxo-1,2,3,4-tetrahydroquinazoline-6-sulfonyl chloride, 2,4-dioxo-1,3-dihydroquinazoline-6-sulfonyl chloride, 6-chlorosulfonyl-1,3-dihydroquinazoline-2,4-dione, pubchem5532, acmc-209lqm, 6-chlorosulfonylquinazoline-2,4-dione, 2,4-dioxo-1,2,3,4-tetrahydro-quinazoline-6-sulfony
PubChem CID 3162025
IUPAC Name 2,4-dioxo-1H-quinazoline-6-sulfonyl chloride
SMILES C1=CC2=C(C=C1S(=O)(=O)Cl)C(=O)NC(=O)N2

Specifications

Quantity 250 mg
UN Number UN3261
Sensitivity Moisture sensitive
Formula Weight 259.97
Percent Purity 97%
Chemical Name or Material 2,4-Dioxo-1,2,3,4-tetrahydroquinazoline-6-sulfonyl chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.