missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Fluorophenoxy)nicotinic acid, 97%, Thermo Scientific™

Product Code. 10713873
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10713873 1 g Amber Glass Bottle 1g
10375302 10 g Amber Glass Bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10713873 Supplier Thermo Scientific Maybridge Supplier No. TL00878DA
 Restricted Item
Estimated Shipment: 25-09-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 54629-13-9
Molecular Formula C12H8FNO3
Molecular Weight (g/mol) 233.198
MDL Number MFCD00833409
InChI Key SDZUYDOXBXHDCE-UHFFFAOYSA-N
Synonym 2-4-fluorophenoxy nicotinic acid, 2-4-fluorophenoxy pyridine-3-carboxylic acid, 2-4-fluoro-phenoxy-nicotinic acid, pubchem16292, acmc-1akqk, maybridge1_000048, 2-4-fluorophenoxy-3-pyridinecarboxylic acid, 3-pyridinecarboxylicacid, 2-4-fluorophenoxy, 3-pyridinecarboxylic acid, 2-4-fluorophenoxy
PubChem CID 605123
IUPAC Name 2-(4-fluorophenoxy)pyridine-3-carboxylic acid
SMILES C1=CC(=C(N=C1)OC2=CC=C(C=C2)F)C(=O)O

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 233.2
Percent Purity 97%
Chemical Name or Material 2-(4-Fluorophenoxy)nicotinic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.