missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Fluorophenyl)benzimidazole, 95%, Thermo Scientific Chemicals

Product Code. 15451789
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15451789 1 g 1g
15461789 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15451789 Supplier Thermo Scientific Alfa Aesar Supplier No. H64531.03
 Restricted Item
Estimated Shipment: 23-09-2026
Add to basket
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 324-27-6
Molecular Formula C13H9FN2
Molecular Weight (g/mol) 212.23
MDL Number MFCD00224358
InChI Key FPWUSPPQEHBWHC-UHFFFAOYSA-N
Synonym 2-4-fluorophenyl-1h-benzimidazole, 1h-benzimidazole, 2-4-fluorophenyl, 2-4-fluorophenyl-1h-1,3-benzodiazole, 2-4-fluorophenyl-1h-benzoimidazole, p-fluorophenyl-2 benzimidazole, 2-4-fluoro-phenyl-1h-benzoimidazole, 2-4-fluorophenyl-1h-benzo d imidazole, p-fluorophenyl-2 benzimidazole french, 2-4-fluorophenyl benzimidazole, benzimidazole, 2-p-fluorophenyl-7ci,8ci
PubChem CID 101259
SMILES FC1=CC=C(C=C1)C1=NC2=CC=CC=C2N1

Specifications

Quantity 1 g
IUPAC Name 2-(4-fluorophenyl)-1H-benzimidazole
Formula Weight 212.23
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 2-(4-Fluorophenyl)benzimidazole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.