missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Fluorophenyl)ethylamine, 97%

Product Code. 15448368
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15448368 100 g 100g
15438368 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15448368 Supplier Thermo Scientific Alfa Aesar Supplier No. H61791.22
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1583-88-6 | C8H10FN | 139.173 g/mol

2-(4-Fluorophenyl)ethylamine may be employed as nucleophile in the synthesis of 2-amino-4-arylpyrimidine derivatives. It is suitable for use in the preparation of ortho-metalated primary phenethylamines having electron-releasing and electron-withdrawing groups on the aromatic ring, leading to complexes containing six membered palladacycles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(4-Fluorophenyl)ethylamine may be employed as nucleophile in the synthesis of 2-amino-4-arylpyrimidine derivatives. It is suitable for use in the preparation of ortho-metalated primary phenethylamines having electron-releasing and electron-withdrawing groups on the aromatic ring, leading to complexes containing six membered palladacycles.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1583-88-6
Molecular Formula C8H10FN
Molecular Weight (g/mol) 139.173
MDL Number MFCD00134208
InChI Key CKLFJWXRWIQYOC-UHFFFAOYSA-N
Synonym 4-fluorophenethylamine, 2-4-fluorophenyl ethanamine, 2-4-fluoro-phenyl-ethylamine, p-fluorophenethylamine, 4-fluorophenylethylamine, 2-4-fluorophenyl ethylamine, 2-4-fluorophenyl ethan-1-amine, benzeneethanamine, 4-fluoro, p-fluorophenyl ethylamine, 2-4-fluorophenyl-ethylamine
PubChem CID 4653
IUPAC Name 2-(4-fluorophenyl)ethanamine
SMILES C1=CC(=CC=C1CCN)F

Specifications

Color Yellow
Density 1.061 g/mL
Boiling Point 50°C to 52°C
Flash Point 79°C (174°F)
Refractive Index 1.507
Quantity 100 g
UN Number UN2922
Solubility Information Slightly soluble in water.
Formula Weight 139.17
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 2-(4-Fluorophenyl)ethylamine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.