missing translation for 'onlineSavingsMsg'
Learn More

2,4-Hexadiene, tech. 90%, mixture of isomers, Thermo Scientific™

Product Code. 15480178
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 15480178

Brand: Thermo Scientific Alfa Aesar H53404.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
2,4-Hexadiene is used in the production of 3,6-dimethyl-3,6-dihydro-[1,2]dithiine.

Solubility
Miscible with ethanol, ethyl ether and chloroform. Immiscible with water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 592-46-1
Molecular Formula C6H10
Molecular Weight (g/mol) 82.15
MDL Number MFCD00009297,MFCD00009297
InChI Key APPOKADJQUIAHP-CIIODKQPSA-N
Synonym unii-g4m9133vs9, 2,4-hexadiene, e,z, cis,trans-2,4-hexadiene, 2,4-hexadiene, e , z-ch3ch=chch=chch3, cis,trans-hexa-2,4-diene, 2,4-hexadiene, 2e,4z, cis-2-trans-4-hexadiene, e,z-2,4-hexadiene, z,e-2,4-hexadiene
PubChem CID 643786
IUPAC Name (2Z,4E)-hexa-2,4-diene
SMILES C\C=C\C=C/C

Specifications

Density 0.72
Boiling Point 82°C
Flash Point −14°C (−7°F)
Linear Formula CH3CH=CHCH=CHCH3
Refractive Index 1.445
Quantity 25 g
UN Number UN2458
Solubility Information Miscible with ethanol,ethyl ether and chloroform. Immiscible with water.
Formula Weight 82.15
Percent Purity 90%
Grade Technical
Chemical Name or Material 2,4-Hexadiene, mixture of isomers
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.