missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Isobutylphenyl)propanoic acid, 98%, BLD Pharm™

Product Code. 30773418
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
1 kg
Unit Size:
100g
1kg
25g
500g
5g
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity missing translation for 'unitSize'
30773418 100 g 100g
30772477 500 g 500g
30764799 1 kg 1kg
30772159 25 g 25g
30767559 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 30773418 missing translation for 'mfr' BLD Pharm missing translation for 'supplierNo' BD134280100g
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 15687-27-1
Molecular Formula C13H18O2
MDL Number MFCD00010393
Quantity 100 g
SMILES C1=CC(=CC=C1C(C(=O)O)C)CC(C)C
Molecular Weight (g/mol) 206.28
Percent Purity 98%
Chemical Name or Material 2-(4-Isobutylphenyl)propanoic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.